missing translation for 'onlineSavingsMsg'
Learn More

N acetyl transferase 5 Antibody (2C6), Novus Biologicals™

Product Code. 18338699 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338699 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338699 Supplier Novus Biologicals Supplier No. H00051126M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

N acetyl transferase 5 Monoclonal antibody specifically detects N acetyl transferase 5 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen N acetyl transferase 5
Applications Western Blot, ELISA, Immunoprecipitation, Sandwich ELISA
Classification Monoclonal
Clone 2C6
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:2000
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH05181
Gene Alias dJ1002M8.1, dJ1175I6.2 (N-terminal acetyltransferase complex ard1 subunit), EC 2.3.1.88, homolog), N(alpha)-acetyltransferase 20, NatB catalytic subunit, N-acetyltransferase 3 homolog, N-acetyltransferase 5, N-acetyltransferase 5 (ARD1 homolog, S. cerevisiae), N-acetyltransferase 5 (GCN5-related, putative), N-acetyltransferase 5, ARD1 subunit (arrest-defective 1, S. cerevisiae, N-alpha-acetyltransferase 20, NatB catalytic subunit, NAT3N-terminal acetyltransferase B complex catalytic subunit NAT5, NAT5dJ1002M8.1 (N-terminal acetyltransferase complex ard1 subunit), NatB complex subunit NAT5, N-terminal acetyltransferase B complex catalytic subunit NAA20, N-terminal acetyltransferase complex ARD1 subunit
Host Species Mouse
Immunogen NAT5 (AAH05181, 1 a.a. ~ 178 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTTLRAFTCDDLFRFNNINLDPLTETYGIPFYLQYLAHWPEYFIVAEAPGGELMGYIMGKAEGSVAREEWHGHVTALSVAPEFRRLGLAAKLMELLEEISERKGGFFVDLFVRVSNQVAVNMYKQLGYSVYRTVIEYYSASNGEPDEDAYDMRKALSRDTEKKSIIPLPHPVRPEDIE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 51126
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.