missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-S-benzyl-L-cysteine, TRC

Product Code. 30133160
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133160 25 mg 25mg
17973025 10 mg 10mg
17993955 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133160 Supplier Toronto Research Chemicals Supplier No. TRCA16842825MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19542-77-9
Molecular Formula C12H15NO3S, C12 H15 N O3 S
Synonym Alanine, N-acetyl-3-(benzylthio)-, L- (8CI)∣N-Acetyl-S-(phenylmethyl)-L-cysteine∣(R)-2-Acetamido-3-(benzylthio)propanoic acid∣Benzylmercapturic acid∣N-Acetyl-S-benzyl-L-cysteine∣N-Acetyl-S-benzylcysteine∣S-Benzyl-(R)-mercapturic acid∣S-Benzyl-N-acetyl-L-cysteine∣S-Benzyl-N-acetylcysteine

Specifications

Format Neat
Molecular Weight (g/mol) 253.32
Product Type Other
MDL Number MFCD00038237
InChI Key BJUXDERNWYKSIQ-NSHDSACASA-N
SMILES CC(=O)N[C@@H](CSCC1=CC=CC=C1)C(O)=O
IUPAC Name (2R)-3-(benzylsulfanyl)-2-acetamidopropanoic acid
Shipping Condition Room Temperature
Purity >95% (HPLC)
Quantity 25 mg
Chemical Name or Material N-Acetyl-S-benzyl-L-cysteine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.