missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-S-benzyl-d5-L-cysteine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-S-benzyl-d5-L-cysteine |
| CAS | 1955496-81-7 |
| Molecular Formula | C12 D5 H10 N O3 S |
| Quantity | 0.05 g |
| Synonym | N-Acetyl-S-benzyl-L-cysteine-D5 (phenyl-D5), N-Acetyl-S-benzyl-d5-L-cysteine, N-Acetyl-S-(phenyl-d5-methyl)-L-cysteine, (2 R)-2-Acetamido-3-(2,3,4,5,6-pentadeuteriobenzylthio)propanoic acid |
| InChI Formula | InChI=1 S/C12H15NO3S/c1-9(14)13-11(12(15)16)8-17-7-10-5-3-2-4-6-10/h2-6,11 H,7-8H2,1H3,(H,13,14)(H,15,16)/t11-/m0/s1/i2D,3 D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(CSC[C@H](NC(=O)C)C(=O)O)c([2 H])c1[2 H] |
| IUPAC Name | (2 R)-2-acetamido-3-[(2,3,4,5,6-pentadeuteriophenyl)methylsulfanyl]propanoic acid |
| Molecular Weight (g/mol) | 258.348 |
| Formula Weight | 258.109 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction