missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-Pepstatin, Thermo Scientific™

Product Code. 15448479
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
This item is not returnable. View return policy

Product Code. 15448479

Brand: Thermo Scientific Alfa Aesar J66917.MB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28575-34-0
Molecular Formula C31H57N5O9
Molecular Weight (g/mol) 643.823
MDL Number MFCD00214071
InChI Key WKYBEGDEGRCZNF-LBTYKNIQSA-N
Synonym acetylpepstatin, acetyl-pepstatin, acetyl pepstatin, ac-val-val-sta-ala-sta, 3s,4s-4-2s-2-3s,4s-4-2s-2-2s-2-acetamido-3-methyl-butanoyl amino-3-methyl-butanoyl amino-3-hydroxy-6-methyl-heptanoyl amino propanoyl amino-3-hydroxy-6-methyl-heptanoic acid, pepstatin ac, pepsidin c, pepsidine c, 5hvp, n-acetyl pepstatin
PubChem CID 5481345
IUPAC Name (3S,4S)-4-[[(2S)-2-[[(3S,4S)-4-[[(2S)-2-[[(2S)-2-acetamido-3-methylbutanoyl]amino]-3-methylbutanoyl]amino]-3-hydroxy-6-methylheptanoyl]amino]propanoyl]amino]-3-hydroxy-6-methylheptanoic acid
SMILES CC(C)CC(C(CC(=O)O)O)NC(=O)C(C)NC(=O)CC(C(CC(C)C)NC(=O)C(C(C)C)NC(=O)C(C(C)C)NC(=O)C)O

Specifications

Color White
Quantity 25 mg
Formula Weight 643.82
Physical Form Solid
Chemical Name or Material N-Acetyl-Pepstatin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.