missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-tyrosine ethyl ester monohydrate, 99%

Product Code. 10785161 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10785161 10 g Glass bottle 10g
10336020 1 g Glass bottle 1g
10761355 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10785161 Supplier Thermo Scientific Acros Supplier No. 102750100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 36546-50-6 | C13H19NO5 | 269.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36546-50-6
Molecular Formula C13H19NO5
Molecular Weight (g/mol) 269.30
MDL Number MFCD00149093
InChI Key YWAVLHZJMWEYTA-HVPILOLGNA-N
Synonym n-acetyl-l-tyrosine ethyl ester monohydrate, ethyl n-acetyl-l-tyrosinate hydrate, ac-tyr-oet.h2o, atee, ethyl 2s-2-acetamido-3-4-hydroxyphenyl propanoate hydrate, n-acetyl-l-tyrosineethylestermonohydrate, n-acetyltyrosine ethyl ester monohydrate, ksc491m4n, n-acetyl tyrosine ethyl ester mono hydrate, acetyl-l-tyrosine ethyl ester hydrate
PubChem CID 2723594
IUPAC Name ethyl (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoate;hydrate
SMILES O.CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 80°C to 81°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula 4-(HO)C6H4CH2CH(NHCOCH3)CO2C2H5·H2O
Quantity 10 g
Specific Rotation + 23
Solubility Information Solubility in water: soluble in cold water. Other solubilities: soluble in hot water,soluble in ethanol
Formula Weight 269.29
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-L-tyrosine ethyl ester monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.