missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-tyrosine ethyl ester monohydrate, 97%

Product Code. 11454643
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454643 5 g 5g
11464643 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454643 Supplier Thermo Scientific Alfa Aesar Supplier No. A11577.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 36546-50-6 | C13H19NO5 | 269.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36546-50-6
Molecular Formula C13H19NO5
Molecular Weight (g/mol) 269.30
MDL Number MFCD00149093
InChI Key YWAVLHZJMWEYTA-HVPILOLGNA-N
Synonym n-acetyl-l-tyrosine ethyl ester monohydrate, ethyl n-acetyl-l-tyrosinate hydrate, ac-tyr-oet.h2o, atee, ethyl 2s-2-acetamido-3-4-hydroxyphenyl propanoate hydrate, n-acetyl-l-tyrosineethylestermonohydrate, n-acetyltyrosine ethyl ester monohydrate, ksc491m4n, n-acetyl tyrosine ethyl ester mono hydrate, acetyl-l-tyrosine ethyl ester hydrate
PubChem CID 2723594
IUPAC Name ethyl (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoate;hydrate
SMILES O.CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O

Specifications

Melting Point 80°C to 81°C
Quantity 5 g
Formula Weight 269.30 (251.28 Anhydrous)
Sensitivity Moisture sensitive
Percent Purity 97%
Chemical Name or Material N-Acetyl-L-tyrosine ethyl ester monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.