missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-methionine-d3 (S-methyl-d3), TRC

Product Code. 30321682
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321682 5 mg 5mg
30309257 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321682 Supplier Toronto Research Chemicals Supplier No. TRCA1863575MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Acetyl-L-Methionine-D3 (S-methyl-D3)
CAS 2714430-55-2
Molecular Formula C7 D3 H10 N O3 S
Quantity 5 mg
Synonym N-Acetyl-L-methionine-d3, N-Acetyl-L-methionine-D3 (S-methyl-D3), (2S)-2-Acetamido-4-(methyl-d3-sulfanyl)butanoic acid
InChI Formula InChI=1S/C7H13NO3S/c1-5(9)8-6(7(10)11)3-4-12-2/h6H,3-4H2,1-2H3,(H,8,9)(H,10,11)/t6-/m0/s1/i2D3
SMILES [2H]C([2H])([2H])SCC[C@H](NC(=O)C)C(=O)O
IUPAC Name (2S)-2-acetamido-4-(trideuteriomethylsulfanyl)butanoic acid
Molecular Weight (g/mol) 194.267
Formula Weight 194.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.