missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-L-glutamic Acid, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-L-glutamic Acid |
| CAS | 1188-37-0 |
| Molecular Formula | C7H11NO5 |
| Quantity | 10 g |
| Synonym | Acetylglutamic Acid, L-N-Acetylglutamic Acid, N-Acetyl-L-glutaminic Acid, N-Acetyl-S-glutamic Acid, N-Acetylglutamic Acid, alpha-(N-Acetyl)-L-glutamic Acid |
| InChI Formula | InChI=1S/C7H11NO5/c1-4(9)8-5(7(12)13)2-3-6(10)11/h5H,2-3H2,1H3,(H,8,9)(H,10,11)(H,12,13)/t5-/m0/s1 |
| SMILES | CC(=O)N[C@@H](CCC(=O)O)C(=O)O |
| IUPAC Name | (2S)-2-acetamidopentanedioic acid |
| Molecular Weight (g/mol) | 189.17 |
| Formula Weight | 189.0637 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction