missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-cysteine, TRC

Product Code. 30134008
Change view
Click to view available options
Quantity:
50 mg
5 g
50 g
Unit Size:
50g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134008 50 g 50g
17993675 50 mg 50mg
17941885 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134008 Supplier Toronto Research Chemicals Supplier No. TRCA17209150G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-91-1
Molecular Formula C5 H9 N O3 S
Molecular Weight (g/mol) 163.19
Synonym N-Acetyl-L-cysteine, (R)-N-Acetylcysteine, ACC, Acetilcysteina, Acetylcysteine, Airbron, Broncholysin, Broncholysin (mucolytic), Brunac, Exomuc, Fabrol, Fluatox, Fluibiotic, Fluimicil, Fluimicil Infantil, Fluimucetin, Fluimucil, Flumil, Fluprowit, Hypotears, L-Acetylcysteine, L-N-Acetylcysteine, Mercapturic acid, (R)-Mercapturic acid, Muco Sanigen, Mucocedyl, Mucofilin, Mucolator, Mucolyticum, Mucolyticum-Lappe, Mucolytikum Lappe, Mucomyst, Mucosolvin, Mucret, N-Acetyl-(R)-cysteine, N-Acetylcysteine, Nα-Acetylcysteine, NSC 111180, Neo-Fluimucil, Parvolex, Respaire, Syntemucol, Tixair
IUPAC Name (2R)-2-acetamido-3-sulfanylpropanoic acid
SMILES CC(=O)N[C@@H](CS)C(=O)O

Specifications

InChI Formula InChI=1S/C5H9NO3S/c1-3(7)6-4(2-10)5(8)9/h4,10H,2H2,1H3,(H,6,7)(H,8,9)/t4-/m0/s1
Quantity 50 g
Formula Weight 163.0303
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material N-Acetyl-L-cysteine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.