missing translation for 'onlineSavingsMsg'
Learn More

N-acetyl-L-cysteine, >96%, MP Biomedicals™

Product Code. 11378399 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378399 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11378399 Supplier MP Biomedicals Supplier No. 0210009850

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 616-91-1
Molecular Formula C5H9NO3S
Molecular Weight (g/mol) 163.191
MDL Number MFCD00004880
InChI Key PWKSKIMOESPYIA-BYPYZUCNSA-N
Synonym n-acetyl-l-cysteine, acetylcysteine, n-acetylcysteine, mercapturic acid, acetadote, l-acetylcysteine, broncholysin, fluimucil, mucomyst, fluprowit
PubChem CID 12035
ChEBI CHEBI:28939
IUPAC Name (2R)-2-acetamido-3-sulfanylpropanoic acid
SMILES CC(=O)NC(CS)C(=O)O

Specifications

Color White
Assay Percent Range >96%
Density 1.295g/cm3 (L)
Quantity 50 g
Formula Weight 163.2
Physical Form Crystalline Powder
Chemical Name or Material N-acetyl-L-cysteine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.