missing translation for 'onlineSavingsMsg'
Learn More

Melford n-Acetyl-L-cysteine

Product Code. 30017932
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
Unit Size:
100g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30017932 500 g 500g
30017931 50 g 50g
30017930 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30017932 Supplier Melford Supplier No. A10040500.0
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Increases cellular pools of free radical scavenger.

Increases cellular pools of free radical scavenger. Prevents apoptotic death of neuronal cells. Induces apoptosis in human and rat smooth muscle cells. Pyrogen free.

Specifications

Chemical Name or Material n-Acetyl-L-cysteine
CAS 616-91-1
Molecular Formula C5H9NO3S
Molecular Weight (g/mol) 163.2
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.