missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-carnosine, TRC

Product Code. 30136428
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136428 10 g 10g
17997654 100 mg 100mg
17977854 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136428 Supplier Toronto Research Chemicals Supplier No. TRCA17201510G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56353-15-2
Molecular Formula C11 H16 N4 O4
Molecular Weight (g/mol) 268.27
IUPAC Name (2S)-2-(3-acetamidopropanoylamino)-3-(1H-imidazol-4-yl)propanoic acid
SMILES CC(=O)NCCC(=O)N[C@@H](Cc1c[nH]cn1)C(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H16N4O4/c1-7(16)13-3-2-10(17)15-9(11(18)19)4-8-5-12-6-14-8/h5-6,9H,2-4H2,1H3,(H,12,14)(H,13,16)(H,15,17)(H,18,19)/t9-/m0/s1
Formula Weight 268.1172
Chemical Name or Material N-Acetyl-L-carnosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.