missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-aspartic Acid, TRC

Product Code. 30008779
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008779 5 g 5g
30137335 25 g 25g
30136962 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008779 Supplier Toronto Research Chemicals Supplier No. TRCA1686055G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 997-55-7
Molecular Formula C6H9NO5
Molecular Weight (g/mol) 175.14
Synonym Acetyl-L-aspartic Acid, Acetylaspartic Acid, L-N-Acetylaspartic Acid, N-Acetyl-L-aspartic Acid, N- Acetyl-S-aspartic Acid, N-Acetylaspartic Acid
IUPAC Name (2S)-2-acetamidobutanedioic acid
SMILES CC(=O)N[C@@H](CC(=O)O)C(=O)O

Specifications

InChI Formula InChI=1S/C6H9NO5/c1-3(8)7-4(6(11)12)2-5(9)10/h4H,2H2,1H3,(H,7,8)(H,9,10)(H,11,12)/t4-/m0/s1
Quantity 5 g
Formula Weight 175.0481
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material N-Acetyl-L-aspartic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.