missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-aspartic acid, MedChemExpress

Product Code. 16435586
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16435586 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16435586 Supplier Medchem Express Supplier No. HY113524
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress N-Acetyl-L-aspartic acid is a derivative of aspartic acid.

N-Acetyl-L-aspartic acid is a derivative of aspartic acid.

Specifications

Chemical Name or Material N-Acetyl-L-aspartic acid
CAS 997-55-7
Color White
Purity Grade Notes Research
Molecular Formula C6H9NO5
Solubility Information H2O : 62.5 mg/mL (356.86 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (28.55 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(O)C[C@@H](C(O)=O)NC(C)=O
Molecular Weight (g/mol) 175.14
Quantity 100 mg
Formula Weight 175.14
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.