missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-Glu-Glu-Val-Val-Pro-Cys-p-nitroanilide, Thermo Scientific™
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spezifikation
Spezifikation
| Chemical Name or Material | N-Acetyl-Glu-Glu-Val-Val-Pro-Cys-p-nitroanilide |
| Color | White |
| MDL Number | MFCD07366698 |
| Quantity | 0.5 mg |
| Synonym | Ac-Glu-Glu-Val-Val-Pro-Cys-pNA |
| Formula Weight | 836.91 |
| Physical Form | Solid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur