missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford n-Acetyl-DL-methionine
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- CAS Number: 115-47-5
- Molecular Formula: C7H13NO3S
- Molecular Weight: 191.3 g/mol
Specifications
Specifications
| Chemical Name or Material | n-Acetyl-DL-methionine |
| CAS | 1115-47-5 |
| Molecular Formula | C7H13NO3S |
| Molecular Weight (g/mol) | 191.3 |
| Quantity | 100 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction