missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-DL-leucine, 99%

Product Code. 11327289
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11327289 25 g 25g
11317289 5 g 5g
11337289 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11327289

Brand: Thermo Scientific Alfa Aesar H27441.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-15-0 | C8H15NO3 | 173.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-15-0
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.21
MDL Number MFCD00026498
InChI Key WXNXCEHXYPACJF-UHFFFAOYNA-N
Synonym n-acetyl-dl-leucine, acetylleucine, ac-dl-leu-oh, tanganil, l-leucine, n-acetyl, acetyl-dl-leucine, acetylleucine inn, dl-leucine, n-acetyl, 2-acetylamino-4-methylpentanoic acid
PubChem CID 1995
IUPAC Name 2-acetamido-4-methylpentanoic acid
SMILES CC(C)CC(NC(C)=O)C(O)=O

Specifications

Melting Point 159°C to 161°C
Quantity 25 g
Formula Weight 173.21
Percent Purity 99%
Chemical Name or Material N-Acetyl-DL-leucine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.