missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-DL-aspartic Acid, TRC

Product Code. 30327206
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327206 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327206 Supplier Toronto Research Chemicals Supplier No. TRCA1679731G
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Acetyl-DL-aspartic Acid
CAS 2545-40-6
Molecular Formula C6 H9 N O5
Quantity 1 g
Synonym Aspartic acid, N-acetyl- (9CI, ACI), Aspartic acid, N-acetyl-, DL- (8CI), N-Acetylaspartic acid (ACI), 2-Acetamidobutanedioic acid, 2-Acetamidosuccinic acid, 2-Acetylamino-succinic acid, N-Acetyl-DL-aspartic acid, NSC 128611
InChI Formula InChI=1S/C6H9NO5/c1-3(8)7-4(6(11)12)2-5(9)10/h4H,2H2,1H3,(H,7,8)(H,9,10)(H,11,12)
SMILES CC(=O)NC(CC(=O)O)C(=O)O
IUPAC Name 2-acetamidobutanedioic acid
Molecular Weight (g/mol) 175.139
Formula Weight 175.048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.