missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-d3-glufosinate H2O, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Glufosinate-N-acetyl-D3 (acetyl-D3) Monohydrate |
| Molecular Formula | C7 D3 H11 N O5 P . H2 O |
| Quantity | 0.05 g |
| Synonym | 2-(Acetyl-d3-amino)-4-(hydroxymethylphosphinyl)butanoic acid monohydrate, 4-[Hydroxy(methyl)phosphoryl]-2-trideuterioacetamidobutanoic acid monohydrate, N-(Acetyl-d3) glufosinate hydrate, N-(Acetyl-d3)-DL-glufosinate monohydrate, Glufosinate-N-acetyl-d3 monohydrate, Glufosinate-N-acetyl-D3 (acetyl-D3) monohydrate |
| InChI Formula | InChI=1 S/C7H14NO5P.H2O/c1-5(9)8-6(7(10)11)3-4-14(2,12)13;/h6H,3-4H2,1-2H3,(H,8,9)(H,10,11)(H,12,13);1H2/i1D3; |
| SMILES | O.[2 H]C([2 H])([2 H])C(=O)NC(CCP(=O)(C)O)C(=O)O |
| IUPAC Name | 4-[hydroxy(methyl)phosphoryl]-2-[(2,2,2-trideuterioacetyl)amino]butanoic acid;hydrate |
| Molecular Weight (g/mol) | 244.197 |
| Formula Weight | 244.09 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction