missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-D-Glucosamine Kinase/NAGK Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-42694G
This item is not returnable.
View return policy
Description
N-Acetyl-D-Glucosamine Kinase/NAGK Polyclonal antibody specifically detects N-Acetyl-D-Glucosamine Kinase/NAGK in Human samples. It is validated for Western Blot
Specifications
| N-Acetyl-D-Glucosamine Kinase/NAGK | |
| Polyclonal | |
| Western Blot | |
| EC 2.7.1.59, GNKGlcNAc kinase, N-acetyl-D-glucosamine kinase, N-acetylglucosamine kinaseHSA242910 | |
| The immunogen for this product maps to a region between residue 50 and 100 of human N-acetylglucosamine kinase using the numbering given in entry NP_060037.2 (GeneID 55577). | |
| 0.1 mL | |
| metabolism | |
| 55577 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction