missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-Glucosamine Kinase/NAGK Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18419423 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419423 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419423 Supplier Novus Biologicals Supplier No. NBP142694AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

N-Acetyl-D-Glucosamine Kinase/NAGK Polyclonal antibody specifically detects N-Acetyl-D-Glucosamine Kinase/NAGK in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen N-Acetyl-D-Glucosamine Kinase/NAGK
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.1.59, GNKGlcNAc kinase, N-acetyl-D-glucosamine kinase, N-acetylglucosamine kinaseHSA242910
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 50 and 100 of human N-acetylglucosamine kinase using the numbering given in entry NP_060037.2 (GeneID 55577).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 55577
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.