missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-D-Glucosamine Kinase/NAGK Antibody (4D12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22554
This item is not returnable.
View return policy
Description
N-Acetyl-D-Glucosamine Kinase/NAGK Monoclonal specifically detects N-Acetyl-D-Glucosamine Kinase/NAGK in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| N-Acetyl-D-Glucosamine Kinase/NAGK | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 2-5ug, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| EC 2.7.1.59, GNKGlcNAc kinase, N-acetyl-D-glucosamine kinase, N-acetylglucosamine kinaseHSA242910 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human NAGK mAb, clone 4D12, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human NAGK protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 4D12 | |
| Unconjugated | |
| NP_060037.2 | |
| NAGK | |
| Recombinant human NAGK (1-344aa) purified from E. coli | |
| 0.1 mL | |
| metabolism | |
| 55577 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction