missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-D-galactosamine

Product Code. 15447329
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447329 500 mg 500mg
15427329 1 g 1g
15437329 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15447329 Supplier Thermo Scientific Chemicals Supplier No. J66095.ME
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1811-31-0 | C8H15NO6 | 221.21 g/mol

N-Acetyl-D-galactosamine is the initial O-linked sugar to many serine and threonine residues in protein glycosylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-D-galactosamine is the initial O-linked sugar to many serine and threonine residues in protein glycosylation.

Solubility
Soluble in water at 50mg/ml

Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1811-31-0
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.21
MDL Number MFCD00136044
InChI Key OVRNDRQMDRJTHS-KEWYIRBNSA-N
Synonym dsstox_cid_28637, dsstox_rid_82907, dsstox_gsid_48711, n-acetyl-, a-d-galactosamine
PubChem CID 60196345
SMILES CC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O

Specifications

Melting Point 160°C
Color White
Quantity 500 mg
Beilstein 2331340
Solubility Information Soluble in water at 50mg/ml
IUPAC Name N-[(2R,3S,4S,5S)-3,4,5,6-tetrahydroxy-1-oxohexan-2-yl]acetamide
Formula Weight 221.21
Physical Form Solid
Chemical Name or Material N-Acetyl-D-galactosamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.