missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-Calicheamicin, MedChemExpress

Product Code. 16470368
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16470368 5 mg 5mg
16441869 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16470368 Supplier Medchem Express Supplier No. HY19791
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress N-Acetyl-Calicheamicin is a potent enediyne antitumor antibiotic.

N-Acetyl-Calicheamicin is a potent enediyne antitumor antibiotic.

Specifications

Chemical Name or Material N-Acetyl-Calicheamicin
Physical Form Solid
CAS 108212-76-6
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C57H76IN3O22S4
For Use With (Application) Cancer-programmed cell death
Synonym N-Acetyl-Calicheamicin γ N-Acetyl-γ-calicheamicin
Solubility Information DMSO : 100 mg/mL (70.90 mM; Need ultrasonic)
SMILES O[C@](CC1=O)(C#C/C=C\C#C2)/C(C([C@H]2O[C@@](O[C@H](C)[C@@H](NO[C@H](O[C@H](C)[C@H]3SC(C(C(C)=C(I)C(O[C@H](O[C@@H](C)[C@H](O)[C@H]4OC)[C@@H]4O)=C5OC)=C5OC)=O)C[C@@H]3O)[C@@H]6O)([H])[C@@H]6O[C@@](OC[C@@H]7N(CC)C(C)=O)([H])C[C@@H]7OC)=C1NC(OC)=O)=C/CSSSC
Molecular Weight (g/mol) 1410.39
Formula Weight 1410.39
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.