missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-Beta-alanine, TRC

Product Code. 30006613
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006613 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30006613 Supplier Toronto Research Chemicals Supplier No. TRCA168070500MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3025-95-4
Molecular Formula C5 H9 N O3
Synonym β-Alanine, N-acetyl-, N-Acetyl-β-alanine, 3-(Acetylamino)propanoic acid, 3-(Acetylamino)propionic acid, 3-Acetamidopropanoic acid, 3-Acetamidopropionic acid, Acetyl-β-alanine, N-Acetyl-3-aminopropanoic acid, NSC 32039
InChI Formula InChI=1S/C5H9NO3/c1-4(7)6-3-2-5(8)9/h2-3H2,1H3,(H,6,7)(H,8,9)
SMILES CC(=O)NCCC(=O)O
IUPAC Name 3-acetamidopropanoic acid
Molecular Weight (g/mol) 131.13
Quantity 500 mg
Formula Weight 131.0582
Shipping Condition Room Temperature
Chemical Name or Material N-Acetyl-beta-alanine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.