missing translation for 'onlineSavingsMsg'
Learn More

N-Acetal Bromopride, TRC

Product Code. 30133873
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133873 50 mg 50mg
17934095 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133873 Supplier Toronto Research Chemicals Supplier No. TRCA17110050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94135-22-5
Molecular Formula C16H24BrN3O3
Molecular Weight (g/mol) 386.28
Synonym 4-(Acetylamino)-5-bromo-N-[2-(diethylamino)ethyl]-2-methoxy-benzamide, Bromopride Impurity A
IUPAC Name 4-acetamido-5-bromo-N-[2-(diethylamino)ethyl]-2-methoxybenzamide
SMILES CCN(CC)CCNC(=O)c1cc(Br)c(NC(=O)C)cc1OC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H24BrN3O3/c1-5-20(6-2)8-7-18-16(22)12-9-13(17)14(19-11(3)21)10-15(12)23-4/h9-10H,5-8H2,1-4H3,(H,18,22)(H,19,21)
Formula Weight 385.1001
Chemical Name or Material N-Acetal Bromopride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.