missing translation for 'onlineSavingsMsg'
Learn More

N-a-Benzoyl-L-arginine Ethyl Ester Hydrochloride, TRC

Product Code. 30347849
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30347849 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30347849 Supplier Toronto Research Chemicals Supplier No. TRCB1220105G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-alpha-Benzoyl-L-arginine Ethyl Ester Hydrochloride
CAS 2645-08-1
Molecular Formula C15H22N4O3.HCl
Quantity 5 g
Synonym Ethyl N-Benzoyl-alpha-arginine Hydrochloride, N-Benzoyl-L-arginine Ethyl Ester Hydrochloride, NSC 86159, alpha-N-Benzoyl-L-Arginine Ethyl Ester Hydrochloride, L-N2-Benzoyl-Arginine Ethyl Ester Monohydrochloride, N2-Benzoyl-L-arginine Ethyl Ester Monohydrochloride
InChI Formula InChI=1S/C15H22N4O3.ClH/c1-2-22-14(21)12(9-6-10-18-15(16)17)19-13(20)11-7-4-3-5-8-11;/h3-5,7-8,12H,2,6,9-10H2,1H3,(H,19,20)(H4,16,17,18);1H/t12-;/m0./s1
SMILES Cl.CCOC(=O)[C@H](CCCNC(=N)N)NC(=O)c1ccccc1
IUPAC Name ethyl (2S)-2-benzamido-5-carbamimidamidopentanoate;hydrochloride
Molecular Weight (g/mol) 342.82
Formula Weight 342.1459
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.