Learn More
N-(9-Fluorenylmethoxycarbonyloxy)succinimide, 98%
Beschreibung
N-(9-Fluorenylmethoxycarbonyloxy)succinimide is the most efficient reagent for the selective preparation of N-(9-fluorenylmethoxycarbonyl) derivatives of hydroxy-amino acids. Dipeptide formation is lower than with the chloroformate, Fmoc-Cl. It is also employed in the synthesis of glycopeptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-(9-Fluorenylmethoxycarbonyloxy)succinimide is the most efficient reagent for the selective preparation of N-(9-fluorenylmethoxycarbonyl) derivatives of hydroxy-amino acids. Dipeptide formation is lower than with the chloroformate, Fmoc-Cl. It is also employed in the synthesis of glycopeptides.
Solubility
Soluble in dimethyl sulfoxide and dimethyl formamide.
Notes
Store in a cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids.
Spezifikation
Spezifikation
| Melting Point | 150°C to 153°C |
| Quantity | 25 g |
| UN Number | UN3077 |
| Beilstein | 3569540 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in dimethyl sulfoxide and dimethyl formamide. |
| Formula Weight | 337.34 |
| Percent Purity | 98% |
| Chemical Name or Material | N-(9-Fluorenylmethoxycarbonyloxy)succinimide |
RUO – Research Use Only
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.