missing translation for 'onlineSavingsMsg'
Learn More

N-(6-Bromohexyl)phthalimide, 97%

Product Code. 11446177
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446177 5 g 5g
11456177 25 g 25g
11466177 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446177 Supplier Thermo Scientific Alfa Aesar Supplier No. B25128.06
 Restricted Item
Estimated Shipment: 21-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 24566-79-8 | C14H16BrNO2 | 310.191 g/mol

N-(6-Bromohexyl)phthalimide is used as a pharmaceutical intermediate and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(6-Bromohexyl)phthalimide is used as a pharmaceutical intermediate and in organic synthesis.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24566-79-8
Molecular Formula C14H16BrNO2
Molecular Weight (g/mol) 310.191
MDL Number MFCD00023098
InChI Key OAZFTIPKNPTDIO-UHFFFAOYSA-N
PubChem CID 141120
IUPAC Name 2-(6-bromohexyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCCCCCBr

Specifications

Melting Point 54°C to 60°C
Boiling Point 186°C to 188°C (0.4 mmHg)
Odor Odorless
Quantity 5 g
Beilstein 186875
Synonym n-6-bromohexyl phthalimide, 2-6-bromohexyl isoindoline-1,3-dione, n-6-bromohexyl-phthalimide, 2-6-bromohexyl-1h-isoindole-1,3 2h-dione, 2-6-bromohexyl isoindole-1,3-dione, 2-6-bromohexyl-2,3-dihydro-1h-isoindole-1,3-dione, 2-6-bromo-hexyl-isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione,2-6-bromohexyl, 1h-isoindole-1,3 2h-dione, 2-6-bromohexyl, 6-n-phthalimido hexyl bromide
Solubility Information Insoluble in water.
Formula Weight 310.19
Percent Purity 97%
Chemical Name or Material N-(6-Bromohexyl)phthalimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.