missing translation for 'onlineSavingsMsg'
Learn More

N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide Hydrochloride, TRC

Product Code. 30314747
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314747 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314747 Supplier Toronto Research Chemicals Supplier No. TRCA61000050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide hydrochloride
CAS 61714-27-0
Molecular Formula C16 H21 Cl N2 O2 S . Cl H
Quantity 50 mg
Synonym W-7 hydrochloride
InChI Formula InChI=1S/C16H21ClN2O2S.ClH/c17-15-9-5-8-14-13(15)7-6-10-16(14)22(20,21)19-12-4-2-1-3-11-18;/h5-10,19H,1-4,11-12,18H2;1H
SMILES Cl.NCCCCCCNS(=O)(=O)c1cccc2c(Cl)cccc12
IUPAC Name N-(6-aminohexyl)-5-chloronaphthalene-1-sulfonamide;hydrochloride
Molecular Weight (g/mol) 377.33
Formula Weight 376.0779
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.