missing translation for 'onlineSavingsMsg'
Learn More

N-(6-Aminohexyl)-1-naphthalenesulfonamide Hydrochloride, TRC

Product Code. 30317405
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30317405 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317405 Supplier Toronto Research Chemicals Supplier No. TRCA61100025MG
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(6-Aminohexyl)-1-naphthalenesulfonamide, Hydrochloride
CAS 61714-25-8
Molecular Formula C16 H22 N2 O2 S . Cl H
Quantity 25 mg
InChI Formula InChI=1S/C16H22N2O2S.ClH/c17-12-5-1-2-6-13-18-21(19,20)16-11-7-9-14-8-3-4-10-15(14)16;/h3-4,7-11,18H,1-2,5-6,12-13,17H2;1H
SMILES Cl.NCCCCCCNS(=O)(=O)c1cccc2ccccc12
IUPAC Name N-(6-aminohexyl)naphthalene-1-sulfonamide;hydrochloride
Molecular Weight (g/mol) 342.88
Formula Weight 342.1169
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.