missing translation for 'onlineSavingsMsg'
Learn More

N-(5-Formylpyrimidin-2-yl)glycine, TRC

Product Code. 30396272
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396272 50 mg
30397263 500 mg
30398692 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396272 Supplier Toronto Research Chemicals Supplier No. TRCF59428850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 915923-65-8
Molecular Formula C7H7N3O3
Molecular Weight (g/mol) 181.14
SMILES OC(=O)CNc1ncc(C=O)cn1

Specifications

InChI Formula InChI=1S/C7H7N3O3/c11-4-5-1-8-7(9-2-5)10-3-6(12)13/h1-2,4H,3H2,(H,12,13)(H,8,9,10)
Quantity 50 mg
Formula Weight 181.0487
Shipping Condition Room Temperature
Chemical Name or Material N-(5-Formylpyrimidin-2-yl)glycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.