missing translation for 'onlineSavingsMsg'
Learn More

N-(5-(1,3-Dioxolan-2-yl)-4-(4-fluorophenyl)-6-isopropylpyrimidin-2-yl)-2-(4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonamido)pyrimidin-5-yl)-2-hydroxy-N-methylethanesulfonamide, TRC

Product Code. 30398123
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398123 25 mg 25mg
30396704 100 mg 100mg
30396847 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398123 Supplier Toronto Research Chemicals Supplier No. TRCF89855525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(5-(1,3-Dioxolan-2-yl)-4-(4-fluorophenyl)-6-isopropylpyrimidin-2-yl)-2-(4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonamido)pyrimidin-5-yl)-2-hydroxy-N-methylethanesulfonamide
Analyte or Component Names N-(5-(1,3-Dioxolan-2-yl)-4-(4-fluorophenyl)-6-isopropylpyrimidin-2-yl)-2-(4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonamido)pyrimidin-5-yl)-2-hydroxy-N-methylethanesulfonamide
Shipping Condition Room Temperature
Molecular Formula C34 H40 F2 N6 O7 S2
InChI Formula InChI=1S/C34H40F2N6O7S2/c1-19(2)28-26(30(21-8-12-23(35)13-9-21)39-33(37-28)41(5)50(7,44)45)25(43)18-51(46,47)42(6)34-38-29(20(3)4)27(32-48-16-17-49-32)31(40-34)22-10-14-24(36)15-11-22/h8-15,19-20,25,32,43H,16-18H2,1-7H3
Solution Type Neat
SMILES CC(C)c1nc(nc(c2ccc(F)cc2)c1C(O)CS(=O)(=O)N(C)c3nc(C(C)C)c(C4OCCO4)c(n3)c5ccc(F)cc5)N(C)S(=O)(=O)C
IUPAC Name N-[5-(1,3-dioxolan-2-yl)-4-(4-fluorophenyl)-6-propan-2-ylpyrimidin-2-yl]-2-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-2-hydroxy-N-methylethanesulfonamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 746.844
Quantity 25 mg
Formula Weight 746.237
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.