missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Vinylbenzyl)-N,N-dimethylamine, 90%, technical, stabilized

Product Code. 10782865 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10782865 5 g Glass bottle 5g
10351501 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10782865 Supplier Thermo Scientific Acros Supplier No. 422550050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2245-52-5 | C11H16N | 162.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2245-52-5
Molecular Formula C11H16N
Molecular Weight (g/mol) 162.26
MDL Number MFCD00078270
InChI Key XQBHAZDVLGNSOJ-UHFFFAOYSA-O
Synonym benzenemethanamine, 4-ethenyl-n,n-dimethyl, n,n-dimethyl-1-4-vinylphenyl methanamine, n-4-vinylbenzyl-n,n-dimethylamine, n,n-dimethyl-4'-vinylbenzylamine, 4-vinylbenzyldimethylamine, p-dimethylaminomethylstyrene, 4-n,n-dimethylaminomethyl styrene, 4-n,n,-dimethylaminomethyl styrene, dimethyl 4-vinylphenyl methyl amine, benzenemethanamine,4-ethenyl-n,n-dimethyl
PubChem CID 519956
IUPAC Name 1-(4-ethenylphenyl)-N,N-dimethylmethanamine
SMILES C[NH+](C)CC1=CC=C(C=C)C=C1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Yellow to Brown
Density 0.9200g/mL
Boiling Point 72°C to 74°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula CH2CHC6H4CH2N(CH3)2
Refractive Index 1.5360 to 1.54
Quantity 5 g
Specific Gravity 0.92
Stabilizer 0.1% P-tert.-butylcatechol
Formula Weight 161.25
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material N-(4-Vinylbenzyl)-N, N-dimethylamine, tech., 90%
Show More Show Less
compliance-icons
Product Identifier
  • N-(4-Vinylbenzyl)-N,N-dimethylamine, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.