missing translation for 'onlineSavingsMsg'
Learn More

N-[4-[(Trifluoroacetyl)amino]benzoyl]-L-glutamic Acid Dimethyl Ester-13C5, TRC

Product Code. 30456891
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456891 25 mg 25mg
30452308 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30456891 Supplier Toronto Research Chemicals Supplier No. TRCT78673325MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[4-[(Trifluoroacetyl)amino]benzoyl]-L-glutamic Acid Dimethyl Ester-13C5
Molecular Formula 13C5 C11 H17 F3 N2 O6
Quantity 25 mg
InChI Formula InChI=1S/C16H17F3N2O6/c1-26-12(22)8-7-11(14(24)27-2)21-13(23)9-3-5-10(6-4-9)20-15(25)16(17,18)19/h3-6,11H,7-8H2,1-2H3,(H,20,25)(H,21,23)/t11-/m0/s1/i7+1,8+1,11+1,12+1,14+1
SMILES CO[13C](=O)[13CH2][13CH2][13C@H](NC(=O)c1ccc(NC(=O)C(F)(F)F)cc1)[13C](=O)OC
IUPAC Name dimethyl (2S)-2-[[4-[(2,2,2-trifluoroacetyl)amino]benzoyl]amino](1,2,3,4,5-13C5)pentanedioate
Molecular Weight (g/mol) 395.274
Formula Weight 395.121
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.