missing translation for 'onlineSavingsMsg'
Learn More

N-[4-(Piperidine-4-carbonyl)phenyl]acetamide, 97%, Thermo Scientific™

Product Code. 10387724
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10387724 10 g Amber Glass Bottle 10g
10082064 250 mg Amber Glass Bottle 250mg
10553055 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10387724 Supplier Thermo Scientific Maybridge Supplier No. MO07288EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 124035-23-0
Molecular Formula C14H18N2O2
Molecular Weight (g/mol) 246.31
MDL Number MFCD08690322
InChI Key LLEYCMVEUNZRIO-UHFFFAOYSA-N
Synonym n-4-piperidine-4-carbonyl phenyl acetamide, n-4-piperidin-4-ylcarbonyl phenyl acetamide, acetamide,n-4-4-piperidinylcarbonyl phenyl, acmc-20acnz, n-4-4-piperidylcarbonyl phenyl acetamide, n-4-4-piperidinyl-carbonyl phenyl-acetamide
PubChem CID 15680074
IUPAC Name N-[4-(piperidine-4-carbonyl)phenyl]acetamide
SMILES CC(=O)NC1=CC=C(C=C1)C(=O)C2CCNCC2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 246.31
Percent Purity 97%
Chemical Name or Material N-[4-(piperidine-4-carbonyl)phenyl]acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.