missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[4-(Piperidine-4-carbonyl)phenyl]acetamide, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 246.31 |
Percent Purity | 97% |
Chemical Name or Material | N-[4-(piperidine-4-carbonyl)phenyl]acetamide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'