missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Piperidin-1-ylbenzyl)cyclopropanamine, ≥95%, Thermo Scientific™

Product Code. 10187762
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10187762 1 g Amber Glass Bottle 1g
10331063 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10187762 Supplier Thermo Scientific Maybridge Supplier No. CC33958DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 857283-76-2
Molecular Formula C15H22N2
Molecular Weight (g/mol) 230.36
MDL Number MFCD07772814
InChI Key UNFZQBXRXGLJLF-UHFFFAOYSA-N
Synonym n-4-piperidin-1-ylbenzyl cyclopropanamine, n-4-piperidin-1-yl phenyl methyl cyclopropanamine, cyclopropyl 4-piperidylphenyl methyl amine, n-4-piperidin-1-yl benzyl cyclopropanamine, benzenemethanamine,n-cyclopropyl-4-1-piperidinyl, n-4-piperidin-1-ylphenyl methyl cyclopropanamine
PubChem CID 7162339
IUPAC Name N-{[4-(piperidin-1-yl)phenyl]methyl}cyclopropanamine
SMILES C(NC1CC1)C1=CC=C(C=C1)N1CCCCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 230.35
Percent Purity 95%
Chemical Name or Material N-(4-piperidin-1-ylbenzyl)cyclopropanamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.