missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Nitrobenzoyl)-L-glutamic acid hemihydrate, 98%, Thermo Scientific™

Product Code. 10741481 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10741481

Brand: Thermo Scientific Acros 205760050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6758-40-3
CAS Min % 97.5
CAS Max % 100.0
Melting Point 111°C to 118°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula 0·5 H2O
Linear Formula HO2CCH2CH2CH(NHCOC6H4NO2)CO2H·1/2H2O
MDL Number MFCD00149508
Quantity 5 g
Beilstein 09, III, 1737
Specific Rotation Condition +15.00 (20.00°C c=2,0.1N NaOH)
Specific Rotation + 15
Synonym s-2-4-nitrobenzamido pentanedioic acid, p-nitrobenzoyl-l-glutamic acid, l-glutamic acid, n-4-nitrobenzoyl, n-p-nitrobenzoyl-l-glutamic acid, n-4-nitrobenzoyl-l-glutamic acid, n-4-nitrobenzoyl-l-glutamic acid hemihydrate, 2s-2-4-nitrophenyl formamido pentanedioic acid, glutamic acid, n-p-nitrobenzoyl-, l, n-4-nitrobenzoyl glutamic acid #
InChI Key NOJZBJAFCSWMKC-VIFPVBQESA-N
SMILES C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)[N+](=O)[O-]
Molecular Weight (g/mol) 305.25
PubChem CID 2724179
Formula Weight 305.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-(4-Nitrobenzoyl)-L-glutamic acid hemihydrate, 98%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.