missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Methoxybenzyl)-N-methylamine, tech, Thermo Scientific™

Product Code. 10082172
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 702-24-9
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
InChI Key AIJFPNKGGAPZFJ-UHFFFAOYSA-N
Synonym 4-methoxy-n-methylbenzylamine, n-4-methoxybenzyl-n-methylamine, n-methyl-4-methoxybenzylamine, 1-4-methoxyphenyl-n-methylmethanamine, 4-methoxyphenyl methyl methyl amine, benzenemethanamine, 4-methoxy-n-methyl, 4-methoxy-benzyl-methyl-amine, 4-methoxyphenyl-n-methylmethanamine, n-methyl-p-methoxybenzylamine, 1-4-methoxyphenyl-n-methyl-methanamine
PubChem CID 485407
IUPAC Name 1-(4-methoxyphenyl)-N-methylmethanamine
SMILES CNCC1=CC=C(C=C1)OC

Specifications

Density 1.0080g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3C6H4CH2NCH3
Quantity 250 mg
Formula Weight 151.21
Percent Purity 97%
Chemical Name or Material N-(4-Methoxybenzyl)-N-methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'