missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Methoxybenzyl)-N-methylamine, tech, Thermo Scientific™

Product Code. 10184542
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10184542 1 g Amber Glass Bottle 1g
10082172 250 mg Amber Glass Bottle 250mg
10633645 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10184542 Supplier Thermo Scientific Maybridge Supplier No. MO00889DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 702-24-9
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
InChI Key AIJFPNKGGAPZFJ-UHFFFAOYSA-N
Synonym 4-methoxy-n-methylbenzylamine, n-4-methoxybenzyl-n-methylamine, n-methyl-4-methoxybenzylamine, 1-4-methoxyphenyl-n-methylmethanamine, 4-methoxyphenyl methyl methyl amine, benzenemethanamine, 4-methoxy-n-methyl, 4-methoxy-benzyl-methyl-amine, 4-methoxyphenyl-n-methylmethanamine, n-methyl-p-methoxybenzylamine, 1-4-methoxyphenyl-n-methyl-methanamine
PubChem CID 485407
IUPAC Name 1-(4-methoxyphenyl)-N-methylmethanamine
SMILES CNCC1=CC=C(C=C1)OC

Specifications

Density 1.0080g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3C6H4CH2NCH3
Quantity 1 g
Formula Weight 151.21
Percent Purity 97%
Chemical Name or Material N-(4-Methoxybenzyl)-N-methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.