missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Isopropylphenyl)-3,5-dinitrobenzamide, 97%

Brand:  Alfa Aesar™ H59066.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415258

  • £358.49 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Isopropylphenyl)-3,5-dinitrobenzamide
329.31
MFCD00583845

Specifications

N-(4-Isopropylphenyl)-3,5-dinitrobenzamide
Solid
329.31
1g
97%
MFCD00583845
Videos
SDS