missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Formylphenyl)methanesulfonamide, TRC

Product Code. 30389635
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389635 5 mg
30395503 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30389635 Supplier Toronto Research Chemicals Supplier No. TRCF7009555MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83922-54-7
Molecular Formula C8 H9 N O3 S
Molecular Weight (g/mol) 199.23
Synonym N-(4-Formylphenyl)-methanesulphonamide, Sotalol Hydrochloride Imp. C (EP)
IUPAC Name N-(4-formylphenyl)methanesulfonamide
SMILES CS(=O)(=O)Nc1ccc(C=O)cc1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C8H9NO3S/c1-13(11,12)9-8-4-2-7(6-10)3-5-8/h2-6,9H,1H3
Formula Weight 199.0303
Chemical Name or Material N-(4-Formylphenyl)-methanesulphonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.