missing translation for 'onlineSavingsMsg'
Learn More

N-[(4-Fluorophenyl)methyl]prop-2-enamide, TRC

Product Code. 30389770
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389770 100 mg -
30390943 10 mg 10mg
30392297 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389770 Supplier Toronto Research Chemicals Supplier No. TRCF594463100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1046471-60-6
Molecular Formula C10H10NOF
Molecular Weight (g/mol) 179.19
SMILES Fc1ccc(CNC(=O)C=C)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H10FNO/c1-2-10(13)12-7-8-3-5-9(11)6-4-8/h2-6H,1,7H2,(H,12,13)
Chemical Name or Material N-[(4-Fluorophenyl)methyl]prop-2-enamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.