missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Diethylaminobenzylideneamino)guanidine Acetic Acid Salt, TRC

Product Code. 30385525
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385525 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385525 Supplier Toronto Research Chemicals Supplier No. TRCD44358010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(4-Diethylaminobenzylideneamino)guanidine Acetic Acid Salt
CAS 1185244-51-2
Molecular Formula C12 H19 N5 . C2 H4 O2
Quantity 10 mg
InChI Formula InChI=1S/C12H19N5.C2H4O2/c1-3-17(4-2)11-7-5-10(6-8-11)9-15-16-12(13)14;1-2(3)4/h5-9H,3-4H2,1-2H3,(H4,13,14,16);1H3,(H,3,4)/b15-9+;
SMILES CCN(CC)c1ccc(\C=N\NC(=N)N)cc1.CC(=O)O
IUPAC Name acetic acid;1-[(E)-[4-(diethylamino)phenyl]methylideneamino]guanidine
Molecular Weight (g/mol) 293.36
Formula Weight 293.1852
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.