missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Chlorophenyl)-N',N'-dimethylurea, 97%, Thermo Scientific™

Product Code. 11353077
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353077 100 g 100g
11343077 25 g 25g
11363077 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11353077 Supplier Thermo Scientific Alfa Aesar Supplier No. L13455.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-68-5
Molecular Formula C9H11ClN2O
Molecular Weight (g/mol) 198.65
MDL Number MFCD00018556
InChI Key BMLIZLVNXIYGCK-UHFFFAOYSA-N
Synonym monuron, 3-4-chlorophenyl-1,1-dimethylurea, chlorfenidim, lirobetarex, monurex, monurox, monuruon, monuuron, telvar, herbicides, monuron
PubChem CID 8800
ChEBI CHEBI:38214
IUPAC Name 3-(4-chlorophenyl)-1,1-dimethylurea
SMILES CN(C)C(=O)NC1=CC=C(Cl)C=C1

Specifications

Melting Point 170°C to 176°C
Density 1.27
Quantity 100 g
UN Number UN3077
Beilstein 2097922
Merck Index 14,6261
Formula Weight 198.65
Percent Purity 97%
Chemical Name or Material N-(4-Chlorophenyl)-N',N'-dimethylurea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.