missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Chloro-6-methoxyphenyl)-2,2-dimethylpropanamide, TRC

Product Code. 30361845
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361845 1 g 1g
30356108 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30361845 Supplier Toronto Research Chemicals Supplier No. TRCC3687701G
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113137-29-4
Molecular Formula C12 H16 Cl N O2
Molecular Weight (g/mol) 241.71
IUPAC Name N-(4-chloro-2-methoxyphenyl)-2,2-dimethylpropanamide
SMILES COc1cc(Cl)ccc1NC(=O)C(C)(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H16ClNO2/c1-12(2,3)11(15)14-9-6-5-8(13)7-10(9)16-4/h5-7H,1-4H3,(H,14,15)
Formula Weight 241.087
Chemical Name or Material N-(4-Chloro-6-methoxyphenyl)-2,2-dimethylpropanamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.