missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Bromophenyl)thiourea, 97%

Product Code. 11377596
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377596 5 g 5g
11367596 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377596 Supplier Alfa Aesar™ Supplier No. L11163.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2646-30-2
Molecular Formula C7H7BrN2S
Molecular Weight (g/mol) 231.11
MDL Number MFCD00041176
InChI Key MRVQULNOKCOGHC-UHFFFAOYSA-N
Synonym 1-4-bromophenyl thiourea, 4-bromophenyl thiourea, n-p-bromophenylthiourea, p-bromophenylthiourea, 1-4-bromophenyl-2-thiourea, thiourea, 4-bromophenyl, n-4-bromophenyl thiourea, urea, 1-p-bromophenyl-2-thio, p-bromophenyl thiourea
PubChem CID 2735620
ChEBI CHEBI:34391
IUPAC Name (4-bromophenyl)thiourea
SMILES NC(=S)NC1=CC=C(Br)C=C1

Specifications

Melting Point 178°C to 184°C
Odor Odorless
Quantity 5 g
UN Number UN2811
Beilstein 2361884
Formula Weight 231.12
Percent Purity 97%
Chemical Name or Material N-(4-Bromophenyl)thiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.