missing translation for 'onlineSavingsMsg'
Learn More

N-[4-(beta-Cyclohexylureidoethyl)benzensulfonyl] N'-Cyclohexylurea, TRC

Product Code. 17934805
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17934805 50 mg 50mg
17904425 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17934805 Supplier Toronto Research Chemicals Supplier No. TRCC98825050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10079-35-3
Molecular Formula C22 H34 N4 O4 S
Molecular Weight (g/mol) 450.59
Synonym Benzenesulfonamide, N-[(cyclohexylamino)carbonyl]-4-[2-[[(cyclohexylamino)carbonyl]amino]ethyl]-, Urea, 1-cyclohexyl-3-[p-[(cyclohexylcarbamoyl)sulfamoyl]phenethyl]- (7CI,8CI), N-[(Cyclohexylamino)carbonyl]-4-[2-[[(cyclohexylamino)carbonyl]amino]ethyl]benzenesulfonamide, N-(Cyclohexylcarbamoyl)-4-[2-[(cyclohexylcarbamoyl)amino]ethyl]benzenesulfonamide, N-(Cyclohexylcarbamoyl)-4-[2-[(cyclohexylcarbamoyl)amino]ethyl]benzenesulphonamide, 1-[[4-[2-[(Cyclohexylcarbamoyl)amino]ethyl]phenyl]sulfonyl]-3-cyclohexylurea, N-[4-(β-Cyclohexylureidoethyl)benzensulfonyl] N'-Cyclohexylurea
IUPAC Name 1-cyclohexyl-3-[4-[2-(cyclohexylcarbamoylamino)ethyl]phenyl]sulfonylurea
SMILES O=C(NCCc1ccc(cc1)S(=O)(=O)NC(=O)NC2CCCCC2)NC3CCCCC3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C22H34N4O4S/c27-21(24-18-7-3-1-4-8-18)23-16-15-17-11-13-20(14-12-17)31(29,30)26-22(28)25-19-9-5-2-6-10-19/h11-14,18-19H,1-10,15-16H2,(H2,23,24,27)(H2,25,26,28)
Formula Weight 450.2301
Chemical Name or Material N-(Cyclohexylcarbamoyl)-4-[2-[(cyclohexylcarbamoyl)amino]ethyl]benzenesulphonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.