missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Aminobenzoyl)-L-glutamic Acid, TRC

Product Code. 30005415
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005415 5 g 5g
17908814 500 mg 500mg
17987734 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005415 Supplier Toronto Research Chemicals Supplier No. TRCA5937705G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4271-30-1
Molecular Formula C12 H14 N2 O5
Molecular Weight (g/mol) 266.25
Synonym (2S)-2-[(4-Aminobenzoyl)amino]pentanedioic Acid, Folic Acid Imp. A (EP), N-(4-Aminobenzoyl)-L-glutamic Acid, Glutamic acid, N-(p-aminobenzoyl)- (6CI,7CI), Glutamic acid, N-(p-aminobenzoyl)-, L- (8CI), (4-Aminobenzoyl)-L-glutamic acid, (p-Aminobenzoyl)-L-glutamic acid, (p-Aminobenzoyl)glutamic acid, N-(p-Aminobenzoyl)-L(+)-glutamic acid, N-(p-Aminobenzoyl)-L-glutamic acid, N-(p-Aminobenzoyl)glutamic acid, NSC 71042
IUPAC Name (2S)-2-[(4-aminobenzoyl)amino]pentanedioic acid
SMILES Nc1ccc(cc1)C(=O)N[C@@H](CCC(=O)O)C(=O)O

Specifications

Analyte or Component Names (2S)-2-[(4-Aminobenzoyl)amino]pentanedioic Acid (N-(4-Aminobenzoyl)-L-glutamic Acid)
Shipping Condition Room Temperature
InChI Formula InChI=1S/C12H14N2O5/c13-8-3-1-7(2-4-8)11(17)14-9(12(18)19)5-6-10(15)16/h1-4,9H,5-6,13H2,(H,14,17)(H,15,16)(H,18,19)/t9-/m0/s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 5 g
Formula Weight 266.0903
Chemical Name or Material (2S)-2-[(4-Aminobenzoyl)amino]pentanedioic Acid (N-(4-Aminobenzoyl)-L-glutamic Acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.